Imprimer

Anti-ORC2 Rat Monoclonal Antibody [clone: 3G6]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB22537-50EA 698 EUR
ABCAAB22537-50
Anti-ORC2 Rat Monoclonal Antibody [clone: 3G6]
Anticorps
Type: Primary
Antigen: ORC2 (origin recognition complex, subunit 2)
Clonality: monoclonal
Clone: 3G6
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR