Imprimer

Anti-GSN Sheep polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB22545-100EA 0 EUR
ABCAAB22545-100
Anti-GSN Sheep polyclonal antibody
Type: Primary
Antigen: GSN (gelsolin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Bovine, Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR