Imprimer

Anti-MHC Class I H2 Dd Mouse monoclonal antibody [clone: D1]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB22571-1EA 550 EUR
ABCAAB22571-1
Anti-MHC Class I H2 Dd Mouse monoclonal antibody [clone: D1]
Type: Primary
Antigen: MHC Class I H2 Dd
Clonality: monoclonal
Clone: D1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR