Imprimer

Anti-CD74 Mouse Monoclonal Antibody [clone: CerCLIP.1]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB22606-100EA 701 EUR
ABCAAB22606-100
Anti-CD74 Mouse Monoclonal Antibody [clone: CerCLIP.1]
Anticorps
Type: Primary
Antigen: CD74 (CD74 molecule, major histocompatibility complex, class II invariant chain)
Clonality: monoclonal
Clone: CerCLIP.1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR