Imprimer

Anti-SELE Rat monoclonal antibody [clone: UZ7]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB22631-100EA 572 EUR
ABCAAB22631-100
Anti-SELE Rat monoclonal antibody [clone: UZ7]
Type: Primary
Antigen: SELE (selectin E)
Clonality: monoclonal
Clone: UZ7
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR