Imprimer

Anti-Rpt5 Mouse monoclonal antibody [clone: TBP1-19]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB22635-50EA 565 EUR
ABCAAB22635-50
Anti-Rpt5 Mouse monoclonal antibody [clone: TBP1-19]
Type: Primary
Antigen: Rpt5
Clonality: monoclonal
Clone: TBP1-19
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR