Imprimer

Anti-PSMC6 Mouse Monoclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB22639-50EA 735 EUR
ABCAAB22639-50
Anti-PSMC6 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: PSMC6 (proteasome (prosome, macropain) 26S subunit, ATPase, 6)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR