Imprimer

Anti-Proteasome 20S beta 6 Mouse monoclonal antibody [clone: MCP421]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB22672-50EA 550 EUR
ABCAAB22672-50
Anti-Proteasome 20S beta 6 Mouse monoclonal antibody [clone: MCP421]
Type: Primary
Antigen: Proteasome 20S beta 6
Clonality: monoclonal
Clone: MCP421
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR