Imprimer

Anti-Cytochrome P450 2B1 + 2B2 Mouse Monoclonal Antibody [clone: b/e3 (9.14)]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB22719-50EA 704 EUR
ABCAAB22719-50
Anti-Cytochrome P450 2B1 + 2B2 Mouse Monoclonal Antibody [clone: b/e3 (9.14)]
Anticorps
Type: Primary
Antigen: Cytochrome P450 2B1 + 2B2
Clonality: monoclonal
Clone: b/e3 (9.14)
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR