Imprimer

Anti-Cytochrome P450 3A1 Mouse Monoclonal Antibody [clone: P6]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB22724-50EA 735 EUR
ABCAAB22724-50
Anti-Cytochrome P450 3A1 Mouse Monoclonal Antibody [clone: P6]
Anticorps
Type: Primary
Antigen: Cytochrome P450 3A1
Clonality: monoclonal
Clone: P6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Pig, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR