Imprimer

Anti-Cytochrome P450 3A2 Sheep polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB22731-50EA 540 EUR
ABCAAB22731-50
Anti-Cytochrome P450 3A2 Sheep polyclonal antibody
Type: Primary
Antigen: Cytochrome P450 3A2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR