Imprimer

Anti-SUMO1 Sheep polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB22738-100EA 603 EUR
ABCAAB22738-100
Anti-SUMO1 Sheep polyclonal antibody
Type: Primary
Antigen: SUMO1 (small ubiquitin-like modifier 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR