Imprimer

Anti-KLF15 Rabbit Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB22851-100EA 697 EUR
ABCAAB22851-100
Anti-KLF15 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: KLF15 (Kruppel-like factor 15)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR