Imprimer

Anti-NFYA Rabbit polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB22886-50EA 0 EUR
ABCAAB22886-50
Anti-NFYA Rabbit polyclonal antibody
Type: Primary
Antigen: NFYA (nuclear transcription factor Y, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR