Imprimer

Anti-TCR V beta 10 Mouse monoclonal antibody [clone: G101]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB23348-125EA 550 EUR
ABCAAB23348-125
Anti-TCR V beta 10 Mouse monoclonal antibody [clone: G101]
Type: Primary
Antigen: TCR V beta 10
Clonality: monoclonal
Clone: G101
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR