Imprimer

Anti-THBS1 Mouse Monoclonal Antibody [clone: A65M]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB23420-100EA 691 EUR
ABCAAB23420-100
Anti-THBS1 Mouse Monoclonal Antibody [clone: A65M]
Anticorps
Type: Primary
Antigen: THBS1 (thrombospondin 1)
Clonality: monoclonal
Clone: A65M
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR