Imprimer

Anti-COL3 Mouse Monoclonal Antibody [clone: 1E7-D7/Col3]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB23445-100EA 933 EUR
ABCAAB23445-100
Anti-COL3 Mouse Monoclonal Antibody [clone: 1E7-D7/Col3]
Anticorps
Type: Primary
Antigen: COL3 (collagen, type III)
Clonality: monoclonal
Clone: 1E7-D7/Col3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR