Imprimer

Anti-CDK4 Rabbit polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB23459-100EA 579 EUR
ABCAAB23459-100
Anti-CDK4 Rabbit polyclonal antibody
Type: Primary
Antigen: CDK4 (cyclin-dependent kinase 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR