Imprimer

Anti-C4A Mouse monoclonal antibody unconjugated [clone: 25.3E3]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB23478-100EA 698 EUR
ABCAAB23478-100
Anti-C4A Mouse monoclonal antibody unconjugated [clone: 25.3E3]
Anticorps
Type: Primary
Antigen: C4A (complement component 4A (Rodgers blood group))
Clonality: monoclonal
Clone: 25300
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR