Imprimer

Anti-Mannan Binding Lectin Mouse Monoclonal Antibody [clone: 6B11]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB23479-100EA 691 EUR
ABCAAB23479-100
Anti-Mannan Binding Lectin Mouse Monoclonal Antibody [clone: 6B11]
Anticorps
Type: Primary
Antigen: Mannan Binding Lectin
Clonality: monoclonal
Clone: 6B11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR