Imprimer

Anti-GC Mouse monoclonal antibody unconjugated [clone: 2G2]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB23480-100EA 691 EUR
ABCAAB23480-100
Anti-GC Mouse monoclonal antibody unconjugated [clone: 2G2]
Anticorps
Type: Primary
Antigen: GC (group-specific component (vitamin D binding protein))
Clonality: monoclonal
Clone: 2G2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR