Imprimer

Anti-MHC Class I alpha Mouse monoclonal antibody [clone: F21-2]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB23483-100EA 540 EUR
ABCAAB23483-100
Anti-MHC Class I alpha Mouse monoclonal antibody [clone: F21-2]
Type: Primary
Antigen: MHC Class I alpha
Clonality: monoclonal
Clone: F21-2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR