Imprimer

Anti-CD81 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: Eat 2]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB23553-50EA 698 EUR
ABCAAB23553-50
Anti-CD81 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: Eat 2]
Anticorps
Type: Primary
Antigen: CD81 (CD81 molecule)
Clonality: monoclonal
Clone: Eat 2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR