Imprimer

Anti-Parainfluenza Virus type 1 Mouse monoclonal antibody [clone: 1231]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB23569-500EA 550 EUR
ABCAAB23569-500
Anti-Parainfluenza Virus type 1 Mouse monoclonal antibody [clone: 1231]
Type: Primary
Antigen: Parainfluenza Virus type 1
Clonality: monoclonal
Clone: 1231
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR