Imprimer

Anti-Interferon gamma Rat Monoclonal Antibody [clone: XMG1.2]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB23637-125EA 698 EUR
ABCAAB23637-125
Anti-Interferon gamma Rat Monoclonal Antibody [clone: XMG1.2]
Anticorps
Type: Primary
Antigen: Interferon gamma
Clonality: monoclonal
Clone: XMG1.2
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR