Imprimer

Anti-ALOX12 Rabbit polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB23678-100EA 596 EUR
ABCAAB23678-100
Anti-ALOX12 Rabbit polyclonal antibody
Type: Primary
Antigen: ALOX12 (arachidonate 12-lipoxygenase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR