Imprimer

Anti-TNFRSF8 Mouse Monoclonal Antibody [clone: CON6D/B5]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB23766-100EA 914 EUR
ABCAAB23766-100
Anti-TNFRSF8 Mouse Monoclonal Antibody [clone: CON6D/B5]
Anticorps
Type: Primary
Antigen: TNFRSF8 (tumor necrosis factor receptor superfamily, member 8)
Clonality: monoclonal
Clone: CON6D/B5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR