Imprimer

Anti-ITGA5 Armenian Hamster monoclonal antibody FITC (Fluorescein) [clone: HM alpha 5]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB23815-50EA 283 EUR
ABCAAB23815-50
Anti-ITGA5 Armenian Hamster monoclonal antibody FITC (Fluorescein) [clone: HM alpha 5]
Type: Primary
Antigen: ITGA5 (integrin, alpha 5 (fibronectin receptor, alpha polypeptide))
Clonality: monoclonal
Clone: HM alpha 5
Conjugation: FITC (Fluorescein)
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR