Imprimer

Anti-THY1 Mouse Monoclonal Antibody [clone: AF-9]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB23894-100EA 733 EUR
ABCAAB23894-100
Anti-THY1 Mouse Monoclonal Antibody [clone: AF-9]
Anticorps
Type: Primary
Antigen: THY1 (Thy-1 cell surface antigen)
Clonality: monoclonal
Clone: AF-9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR