Imprimer

Anti-Semaphorin 4D Mouse monoclonal antibody [clone: 133.1G6]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB23899-100EA 572 EUR
ABCAAB23899-100
Anti-Semaphorin 4D Mouse monoclonal antibody [clone: 133.1G6]
Type: Primary
Antigen: Semaphorin 4D
Clonality: monoclonal
Clone: 133.1G6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR