Imprimer

Anti-Dynein intermediate chain 1 Mouse Monoclonal Antibody [clone: 74.1]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB23905-100EA 750 EUR
ABCAAB23905-100
Anti-Dynein intermediate chain 1 Mouse Monoclonal Antibody [clone: 74.1]
Anticorps
Type: Primary
Antigen: Dynein intermediate chain 1
Clonality: monoclonal
Clone: 74.1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR