Imprimer
Anti-FNBP1 Goat polyclonal antibody unconjugated
Fournisseur: Abcam
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ABCAAB23934-100EA 658 EUR
ABCAAB23934-100
Anti-FNBP1 Goat polyclonal antibody unconjugated
Anticorps
Type: Primary
Antigen: FNBP1 (formin binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human