Imprimer

Anti-INCENP Mouse Monoclonal Antibody [clone: 58-217]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB23956-100EA 729 EUR
ABCAAB23956-100
Anti-INCENP Mouse Monoclonal Antibody [clone: 58-217]
Anticorps
Type: Primary
Antigen: INCENP (inner centromere protein antigens 135/155kDa)
Clonality: monoclonal
Clone: 58-217
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR