Imprimer

Anti-Verotoxin II / SLT 2a Mouse monoclonal antibody [clone: 7585]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB23987-50EA 550 EUR
ABCAAB23987-50
Anti-Verotoxin II / SLT 2a Mouse monoclonal antibody [clone: 7585]
Type: Primary
Antigen: Verotoxin II / SLT 2a
Clonality: monoclonal
Clone: 7585
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR