Imprimer

Anti-MHC Class II Mouse Monoclonal Antibody [clone: MRC OX-6]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB23990-100EA 729 EUR
ABCAAB23990-100
Anti-MHC Class II Mouse Monoclonal Antibody [clone: MRC OX-6]
Anticorps
Type: Primary
Antigen: MHC Class II
Clonality: monoclonal
Clone: MRC OX-6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR