Imprimer

Anti-CD8A Mouse monoclonal antibody FITC (Fluorescein) [clone: OX-8]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB23994-100EA 684 EUR
ABCAAB23994-100
Anti-CD8A Mouse monoclonal antibody FITC (Fluorescein) [clone: OX-8]
Type: Primary
Antigen: CD8A (CD8a molecule)
Clonality: monoclonal
Clone: OX-8
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR