Imprimer

Anti-CD1A Mouse Monoclonal Antibody [clone: NA1/34-HLK]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB24055-100EA 704 EUR
ABCAAB24055-100
Anti-CD1A Mouse Monoclonal Antibody [clone: NA1/34-HLK]
Anticorps
Type: Primary
Antigen: CD1A (CD1a molecule)
Clonality: monoclonal
Clone: NA1/34-HLK
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR