Imprimer

Anti-Adenovirus Goat polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB24155-100EA 589 EUR
ABCAAB24155-100
Anti-Adenovirus Goat polyclonal antibody
Type: Primary
Antigen: Adenovirus
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR