Imprimer

Anti-HRP Rabbit polyclonal antibody Biotin

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB24202-500EA 550 EUR
ABCAAB24202-500
Anti-HRP Rabbit polyclonal antibody Biotin
Type: Primary
Antigen: HRP
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR