Imprimer

Anti-La Crosse Virus G1 attachment glycoprotein Mouse monoclonal antibody [clone: 8C2.2]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB24296-50EA 565 EUR
ABCAAB24296-50
Anti-La Crosse Virus G1 attachment glycoprotein Mouse monoclonal antibody [clone: 8C2.2]
Type: Primary
Antigen: La Crosse Virus G1 attachment glycoprotein
Clonality: monoclonal
Clone: 8C2.2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR