Imprimer

Anti-Interferon beta Mouse Monoclonal Antibody [clone: B-02]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB24309-100EA 691 EUR
ABCAAB24309-100
Anti-Interferon beta Mouse Monoclonal Antibody [clone: B-02]
Anticorps
Type: Primary
Antigen: Interferon beta
Clonality: monoclonal
Clone: B-02
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR