Imprimer
Anti-NR1H2 Goat Polyclonal Antibody
Fournisseur: Abcam
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ABCAAB24361-100EA 704 EUR
ABCAAB24361-100
Anti-NR1H2 Goat Polyclonal Antibody
Anticorps
Type: Primary
Antigen: NR1H2 (nuclear receptor subfamily 1, group H, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human