Imprimer

Anti-EFHD2 Goat polyclonal antibody unconjugated

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB24368-100EA 721 EUR
ABCAAB24368-100
Anti-EFHD2 Goat polyclonal antibody unconjugated
Anticorps
Type: Primary
Antigen: EFHD2 (EF-hand domain family, member D2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR