Imprimer

Anti-NTF3 Sheep polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB24494-100EA 572 EUR
ABCAAB24494-100
Anti-NTF3 Sheep polyclonal antibody
Type: Primary
Antigen: NTF3 (neurotrophin 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR