Imprimer

Anti-NEFM Chicken Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB24522-50EA 691 EUR
ABCAAB24522-50
Anti-NEFM Chicken Polyclonal Antibody
Anticorps
Type: Primary
Antigen: NEFM (neurofilament, medium polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR