Imprimer

Anti-TBL1X Rabbit Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB24548-100EA 744 EUR
ABCAAB24548-100
Anti-TBL1X Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: TBL1X (transducin (beta)-like 1X-linked)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR