Imprimer

Anti-PECAM1 Mouse Monoclonal Antibody [clone: P2B1]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB24590-100EA 759 EUR
ABCAAB24590-100
Anti-PECAM1 Mouse Monoclonal Antibody [clone: P2B1]
Anticorps
Type: Primary
Antigen: PECAM1 (platelet/endothelial cell adhesion molecule 1)
Clonality: monoclonal
Clone: P2B1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR