Imprimer

Anti-TUBA Mouse Monoclonal Antibody [clone: 6-11B-1]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB24610-50EA 749 EUR
ABCAAB24610-50
Anti-TUBA Mouse Monoclonal Antibody [clone: 6-11B-1]
Anticorps
Type: Primary
Antigen: TUBA (tubulin, alpha)
Clonality: monoclonal
Clone: 6-11B-1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR