Imprimer

Anti-KRT1 Rabbit polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB24643-50EA 599 EUR
ABCAAB24643-50
Anti-KRT1 Rabbit polyclonal antibody
Type: Primary
Antigen: KRT1 (keratin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR