Imprimer

Anti-NSE Mouse Monoclonal Antibody [clone: NSE-P1]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB24709-100EA 704 EUR
ABCAAB24709-100
Anti-NSE Mouse Monoclonal Antibody [clone: NSE-P1]
Anticorps
Type: Primary
Antigen: NSE
Clonality: monoclonal
Clone: NSE-P1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR